Would you like to log in again?
Blue Phantom® Breast Biopsy Ultrasound Trainer
Supplier: ELEVATE HEALTHCARE, INC. SE
Blue Phantom®
BPB170
470327-322EA 717.19 USD InStock
470327-322
Blue Phantom® Breast Biopsy Ultrasound Trainer
Models Human Biology Models
Save money with durable models.
  • Feels and biopsies like human tissue
  • Hyperechoic, hypoechoic, and echolucent masses
  • Masses from 4 to 11 mm
  • Self-healing tissue
Blue Phantom®'s breast ultrasound medical training model allows users to develop and practice the skills necessary to gain proficiency in using ultrasound for imaging and surgical procedures. Using Blue Phantom® proprietary simulated human tissue, this very realistic and ultra-durable breast ultrasound surgical training model is excellent for training clinicians in the psychomotor skills associated with breast ultrasound guided fine needle biopsy procedures. These ultrasound imaging skills include; using ultrasound system controls, transducer positioning and movement, recognition of soft tissue masses, breast lesions, and breast tumors in human soft tissue, and using ultrasound to target the lesions for fine needle biopsy.
This medical ultrasound training phantom is constructed using Blue Phantom®'s patented ultra-durable tissue and is extremely realistic in ultrasound imaging characteristics and feels like real human tissue. Blue Phantom® breast ultrasound phantoms contain a variety of masses that are hyperechoic, hypoechoic, and echolucent allowing users to gain experience utilizing a wide range of lesions as they would experience in the clinical environment. Each model contains 14 masses of varying sizes (ranging from 4 to 11mm) allowing users to develop their skills starting with larger lesions and target smaller masses as their skills progress.
Ordering information: These items cannot be returned if used. Please consult with your sales representitive before purchasing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty