Would you like to log in again?
Blue Phantom® Full Body FAST Ultrasound Torso Trainer
Supplier: ELEVATE HEALTHCARE, INC. SE
Blue Phantom®
BPFC1704 BPFAST1800 BPFAST1802
470327-346EA 39303.39 USD InStock
470327-346 470327-344 470327-462
Blue Phantom® Full Body FAST Ultrasound Torso Trainer
Models Human Biology Models
Use your ultrasound system to train users to perform a fast exam.
  • Available with transesophageal echocardiography (TEE) option
  • Use any ultrasound imaging system
  • High quality
  • Extremely durable; use for repeated training
Use your ultrasound system to train users to perform FAST exams, TTE, and Pericardiocentesis procedures. Constructed using Blue Phantom®’s patented simulated human tissue, users will experience superb ultrasound imaging characteristics of the structures in the thorax, right upper quadrant, left upper quadrant, abdomen, and pelvis while encountering the same imaging challenges as found in a human patient, such as applying the adequate transducer pressure in order to obtain images, bowel gas, and intercostal access.
Blue Phantom® expends tremendous effort to match the imaging characteristics and feel of real human tissue, allowing teaching and training under optimal conditions. The best simulation training scenarios are requisite on users experiencing the same procedural realism as performing FAST exam, TTE, and Pericardiocentesis procedures and on live patients. This realism makes training better: all structures match the sonographic acoustic characteristics of real human tissue providing uncompromising image quality.
Adjustable internal bleeding levels allow users to individually vary the training scenarios based on the scenarios being presented. This fully imageable upper and lower torso offers extremely realistic internal bleeding in each organ space that can be adjusted by the user to simulate a wide variety of effusion states including: small, medium, and large effusions or no effusions at all around the liver, spleen, heart, and bladder.
Ordering information: These items cannot be returned if used. Please consult with your sales representitive before purchasing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty