Would you like to log in again?
Blue Phantom® Musculoskeletal Knee Ultrasound Trainer
Supplier: ELEVATE HEALTHCARE, INC. SE
Blue Phantom®
BPMSK-1000
470327-402EA 4946.19 USD InStock
470327-402
Blue Phantom® Musculoskeletal Knee Ultrasound Trainer
Models Human Biology Models
Constructed from a digital human file.
  • Superb image quality
  • Use with any ultrasound system
  • Self-healing tissue
  • Adjustable volume in the Bursa and Suprapatellar Space to simulate various injections and aspirations
  • Lightweight and easy to transport
Blue Phantom® is proud to introduce the World's first musculoskeletal ultrasound training model for joint injections of the knee. Constructed utilizing our innovative Life Cast™ modeling approach, the external landmarks were cast from a human knee while the internal anatomy is based on digital human files. This ultra-durable ultrasound simulator incorporates all of the anatomy required to teach, learn, and practice the skills associated with MSK joint injections and aspirations of the knee and was designed for both ultrasound guided and blind insertion procedural training. Utilizing advancements in SimulexUS™ technology we are able to accurately replicate the tendons and ligaments of the knee to offer a truly lifelike training environment, and because SimulexUS™ tissue is self-healing, the need to purchase replacement parts is dramatically reduced.
Ordering information: These items cannot be returned if used. Please consult with your sales representitive before purchasing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty