Would you like to log in again?
Blue Phantom® PUBS Ultrasound Trainer
Supplier: ELEVATE HEALTHCARE, INC. SE
Blue Phantom®
BP1620
470327-414EA 9563.09 USD InStock
470327-414
Blue Phantom® PUBS Ultrasound Trainer
Models Human Biology Models
Superb image quality.
  • Highly realistic
  • Syringe actuated arterial pulsation
  • Easy to refill
  • Use with any ultrasound system
Blue Phantom®'s new percutaneous umbilical cord blood sampling (PUBS) - and Fetal Umbilical Blood Sampling (FUBS) ultrasound training model is an excellent tissue simulation trainer for ultrasound guided procedure training. Built as an extension on our amniocentesis ultrasound trainer, this hands-on training model contains an 18-week fetus with umbilical cord and offers the user an excellent skills development and assessment tool for teaching or learning the complete cordocentesis procedure.
Ordering information: These items cannot be returned if used. Please consult with your sales representitive before purchasing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty