Would you like to log in again?
Blue Phantom® True Anatomy Leg Ultrasound Trainer
Supplier: ELEVATE HEALTHCARE, INC. SE
Blue Phantom®
BPLI410 BPLI405 BPLI405-DVT BPL600 BPL400 BPLI420 BPL400-DVT BPL550
470327-554EA 3705.39 USD InStock
470327-554 470327-454 470327-552 470327-548 470327-458 470327-452 470327-550 470327-450
Blue Phantom® True Anatomy Leg Ultrasound Trainer
Models Human Biology Models
Extremely durable; use for repeated training.
  • Self-healing tissue
  • Use any ultrasound system
  • Matches acoustic characteristics of real tissue
Blue Phantom® leg models are excellent for training clinicians in the techniques associated with ultrasound guided treatment. Aiding users in the development of the psycho-motor skills necessary to gain proficiency in using ultrasound, these ultrasound phantoms contain a wide variety of objects with distinctive sonographic characteristics including metal projectile BBs, soft tissue masses, and vascular conditions. Utilized to develop ultrasound imaging skills as using ultrasound system controls, transducer positioning and movement, recognition of a wide variety of foreign bodies in soft tissue, targeting the foreign bodies, tissue biopsies, and vascular treatment.
Constructed using Blue Phantom® patented ultra-durable tissue, this mannequin is extremely realistic in ultrasound imaging characteristics and feels like real human tissue. This foreign body medical ultrasound training model contains a wide variety of foreign bodies dispersed throughout the model at different depths and positions. The Soft tissue mass model contains 20 masses of varying sizes (ranging from 4 to 11 mm) allowing users to develop their skills starting with larger lesions and target smaller masses as their skills progress.
The vascular and saphenofemoral access models can be purchased as either standard, or containing a deep vein thrombosis (DVT). For the best value, the Sclerotherapy kit includes the base model and three inserts with varying vessel anatomy for progressively more difficult training. All Inserts are interchangeable, making it easy to upgrade to new options.
Ordering information: These items cannot be returned if used. Please consult with your sales representitive before purchasing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty