Would you like to log in again?
Blue Phantom® Lumbar Puncture Ultrasound Trainers
Supplier: ELEVATE HEALTHCARE, INC. SE
Blue Phantom®
BPLP2203 BPLP2102 BPLP2101 BPLP2201 BPLP2103 BPLP2204 BPLP2105 BPLP2202
470327-478EA 2390.59 USD InStock
470327-478 470327-434 470327-466 470327-432 470327-558 470327-436 470327-562 470327-584
Blue Phantom® Lumbar Puncture Ultrasound Trainers
Models Human Biology Models
Available in a variety of configurations to meet your training needs.
  • Extremely durable and self-healing
  • Identify optimal insertion points, angle of needle insertion, and depth to the Ligamentum flavum, epidural space, and spinal cistern
  • Superb ultrasound imaging characteristics
  • Use any ultrasound system
  • Use in upright or lateral position
Blue Phantom® Lumbar puncture and spinal epidural training model provides clinicians the flexibility to practice and teach a wide variety of procedures including lumbar puncture, lumbar epidural, thoracic epidural, and cervical epidural diagnostic and therapeutic procedures. This Lumbar puncture trainer with lumbar epidural is available in a variety of configurations to meet your training needs. The base model contains the lumbar puncture and lumbar epidural configuration. Users are able to add an additional spinal region to the base platform at an additional cost. These optional configurations include thoracic epidural and cervical epidural modules.
Ordering information: These items cannot be returned if used. Please consult with your sales representitive before purchasing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty