Would you like to log in again?
Blue Phantom® Fluids For Ultrasound Training
Supplier: ELEVATE HEALTHCARE, INC. SE
Blue Phantom®
BPP-802 BRS182-CLEAR-1LTR BRS182-CLEAR-235ML BRS180-RED-235ML BRS183-YELOW-1LTR BRS180-RED-1LTR BPP-803 BRS183-YELOW-235ML BRS185-DOPPLER
470327-566EA 56.39 USD InStock
470327-566 470327-168 470327-480 470327-100 470327-166 470327-594 470327-098 470327-482 470327-592
Blue Phantom® Fluids For Ultrasound Training
Models Human Biology Models
Simulate body fluids in ultrasound training.
  • Accurate acoustic properties for blood serum
  • Same viscosity as blood serum
  • Long life; contains preservatives that inhibit bacterial and fungal growth
  • Easy to refill
These simulated blood refills are designed to mimic blood and body fluids in the Blue Phantom® models. Blue and Red simulated blood refill fluid is specifically designed to supplement or replace the venous and arterial blood in Blue Phantom® models containing arteries and veins. Clear replacement fluid is specifically designed to supplement or replace the fluid found in Blue Phantom® spinal epidural and lumbar puncture, paracentesis and amniocentesis ultrasound training model mannequins. Yellow replacement fluid is specifically designed as replacement fluid for Blue Phantom® thoracentesis ultrasound training model. Red Doppler is designed for use with the Blue Phantom® doppler models.
Maintaining the acoustic properties that accurately match the properties found in blood serum, these simulated blood replacement fluids contain bacterial and fungal growth inhibitors so that the fluid can be placed into the model for extended use. As users access the simulated vessels with needles and catheters during simulation training, fluid is often removed and discarded, thus requiring that the blood vessels be replenished with simulated blood. Air might also be introduced into the vessels during training. The amount of simulated blood used is directly dependent upon the amount of fluid removed during simulation training. The phantom's fluid can be easily refilled and air can be purged from the system without difficulty.
This replacement fluid must be utilized, rather than user formulated fluid, in order to maintain the product warranty on the Blue Phantom® and CAE® Products.
Ordering information: These items cannot be returned if used. Please consult with your sales representitive before purchasing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty