Would you like to log in again?
Blue Phantom® Gen II PICC, IV, and Arterial Line Ultrasound Trainer
Supplier: CAE HEALTHCARE, INC. SE
Blue Phantom®
BPA304-HP BPAI305
470327-534EA 1434.39 USD InStock
470327-534 470327-392
Blue Phantom® Gen II PICC, IV, and Arterial Line Ultrasound Trainer
Models Human Biology Models
Confirm placement using ultrasound or X-ray.
  • Self-healing tissue
  • Matches acoustic properties of real tissue
  • Use with any ultrasound system
  • Pulsating arteries via included hand-bulb
  • User replaceable tissue inserts
New to the second generation PICC with I.V. and arterial ultrasound training model is the inclusion of an ultrasoundable chest that contains the superior vena cava and subclavian vein which is designed to allow users to verify correct catheter placement. In addition to ultrasound, correct catheter placement can also be verified using X-ray.
Using Blue Phantom® proprietary simulated human tissue, this very realistic and ultra-durable venous access medical model is excellent for training clinicians in the psychomotor skills associated with ultrasound guided venous and arterial access procedures. These ultrasound imaging skills include; using ultrasound system controls, transducer positioning and movement, recognition of arterial and venous anatomy, using ultrasound to target the appropriate vessels for cannulation, and using ultrasound to guide venous access and arterial access procedures in the arm.
Ordering information: These items cannot be returned if used. Please consult with your sales representitive before purchasing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty