Would you like to log in again?
Blue Phantom® Central Line Ultrasound Trainers
Supplier: ELEVATE HEALTHCARE, INC. SE
Blue Phantom®
BPH660-AP BPH663 BPH665-C-HP BPH667-C BPH660-HP BPH665-C-AP BPH662-B
470327-536EA 3227.39 USD InStock
470327-536 470327-382 470327-470 470327-380 470327-538 470327-390 470327-394
Blue Phantom® Central Line Ultrasound Trainers
Models Human Biology Models
Superior realism, reliability, and durability.
  • Self-healing tissue offers users tremendous durability
  • Anatomically correct; constructed from a digital human file
  • Veins are compressible while the arteries remain uncompressed
  • Easy to refill vessels using Quick fill™ port
Blue Phantom® is proud to introduce the most realistic ultrasound central line mannequin available anywhere. Constructed utilizing the innovative Life Cast™ modeling approach, the external landmarks were cast from a live human while the internal anatomy is based on digital human files. This ultra-durable ultrasound simulator incorporates all of the anatomy required to teach, learn, and practice the skills associated with central line placement and was designed for both ultrasound guided and blind insertion procedural training.
The self-healing tissue withstands tremendous use, minimizing the need for replacement parts. Blue Phantom®'s new upper torso central venous catheter model offers clinicians the most life-like training environment at a low cost of ownership. Perform complete central line placements of the internal jugular and subclavian vein - including needles, guidewires, dilation, and threading of catheters. On all models arterial pulsations are provided using a hand pump, or an integrated automatic pump.
Ordering information: These items cannot be returned if used. Please consult with your sales representitive before purchasing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty