Would you like to log in again?
Blue Phantom® Midscapular Thoracentesis Ultrasound Trainer
Supplier: ELEVATE HEALTHCARE, INC. SE
Blue Phantom®
BPTT2-1005 BPTT2-1006
470327-400EA 7028.59 USD InStock
470327-400 470327-560
Blue Phantom® Midscapular Thoracentesis Ultrasound Trainer
Models Human Biology Models
Practice using ultrasound system controls.
  • Positive fluid flow provides immediate feedback
  • Matches acoustic characteristics of real tissue
  • Use any ultrasound imaging system
  • Extremely durable; use for repeated training
Blue Phantom®'s mid-scapular thoracentesis ultrasound training model is specifically designed for ultrasound guided thoracentesis procedures. This ultrasound training phantom aids users in developing and practicing the skills associated with a mid scapulary approach to ultrasound guided thoracentesis procedures. This model is excellent for assisting clinicians in gaining proficiency in using ultrasound to identify and guide needle and small gauge catheter insertions in a patient with pleural effusions for diagnostic and/or therapeutic purposes.
The model is of a supine adult male and extends from the upper buttocks to the lower neck. Positioned in the mid scapulary line, the ultrasound tissue insert contains chest wall superficial tissue, 6th, 7th, 8th, and 9th ribs and intercostal spaces, pleural cavity with lung and atelectatic lung, diaphragm, and superior spleen. The pleural fluid collections allow users to develop and refine their ultrasound guided thoracentesis skills. This model is extremely realistic and is self-healing offering you superb training with a low cost of ownership.
The ultrasound tissue insert offers extremely realistic sonographic imaging characteristics is designed for guiding the placement of needles and small catheters (18-21 gauge and associated catheter kits). Blue Phantom® simulated human tissue is very realistic and ultra-durable; excellent for repeated training in skills associated with ultrasound guided thoracentesis procedures. Positive fluid flow offers users feedback when pleural effusions are accurately accessed. The fluid is easily refilled using a quick fill luer lock or can be connected to an I.V. reservoir for continuous fluid delivery. Users can also learn to avoid accessory structures as the spleen, diaphragm, and lung.
Ordering information: These items cannot be returned if used. Please consult with your sales representitive before purchasing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty