Would you like to log in again?
Blue Phantom® Renal Biopsy Ultrasound Trainer
Supplier: ELEVATE HEALTHCARE, INC. SE
Blue Phantom®
BPRB2011 BPRB2012 BPRB2013
470327-568EA 717.39 USD InStock
470327-568 470327-418 470327-570
Blue Phantom® Renal Biopsy Ultrasound Trainer
Models Human Biology Models
Excellent imaging characteristics.
  • Anatomically correct adult torso
  • Superb kidney architecture
  • Ultra-durable self-healing tissue
  • Matches the acoustic characteristics of real human tissue
  • Use any ultrasound system
Blue Phantom® renal biopsy ultrasound training models provides users extremely realistic sonographic imaging characteristics and feels like real human tissue. Blue Phantom®'s patented ultra-durable self-healing tissue provides users with a low cost of ownership. In addition to training users how to utilize ultrasound to image the kidney, users are able to perform two types of procedures on the model core needle biopsy procedures and needle aspiration procedures.
Ultrasound imaging and procedural skills include; using ultrasound system controls, transducer positioning and movement, recognition of kidney anatomy, the appearance of ribs and avoiding them during renal biopsy, using ultrasound to target the appropriate location for renal biopsy, and performing an ultrasound guided renal biopsy procedure. This ultrasound hands-on training model will perform well using any ultrasound imaging system configured with the appropriate renal biopsy transducer.
This ultrasound phantom model is excellent for specialties including nephrology, interventional radiology, surgery, surgical training programs, ultrasound training programs, simulation centers, surgical skills centers, medical education facilities, renal biopsy device manufacturers, and ultrasound manufacturers for ultrasound education and demonstrations.
Ordering information: These items cannot be returned if used. Please consult with your sales representitive before purchasing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty