Would you like to log in again?
Blue Phantom® Tap Block Regional Anesthesia Ultrasound Trainer
Supplier: ELEVATE HEALTHCARE, INC. SE
Blue Phantom®
BP2002-TAP BP2001-TAP
470327-576EA 3466.69 USD InStock
470327-576 470327-438
Blue Phantom® Tap Block Regional Anesthesia Ultrasound Trainer
Models Human Biology Models
Utilize traditional anatomical landmarks or guided ultrasound.
  • Provides tactile feedback through the tissue layers
  • Superb imaging characteristics
  • Self-healing tissue will withstand tremendous use
  • Use any ultrasound imaging system
Blue Phantom®'s TAP block ultrasound training model assists users in the development of the psychomotor skills associated with performing ultrasound guided transversus abdominis plane (TAP) block regional anesthesia procedures. Built on Blue Phantom®'s superb realism and unmatched durability of synthetic tissues, the TAP block ultrasound training model assists users in the identification, localization, and targeting of structures utilized in performing this regional anesthesia procedure.
These structures include the cutaneous and subcutaneous tissues, external oblique muscle, internal oblique muscle, transversus abdominis muscles, and the peritoneum. Users are able to simulate the entire TAP block procedure using this highly realistic and durable ultrasound training model. With the goal of depositing local anesthetic in the plane between the internal oblique and transversus abdominis muscles, users can inject simulated anesthetic into the model repeatedly as the fluid is automatically expelled.
We know that gaining proficiency in using ultrasound requires practice. The self-healing tissue will allow you to repeatedly perform TAP block procedures without giving second thought to wearing out the tissue or calculating the cost per cannulation. This model is excellent for teaching a variety of skills including; using ultrasound system controls, transducer positioning and movement, recognition of the subcutaneous tissues, external oblique muscle, internal oblique muscle, transversus abdominis muscles, and the peritoneal border, using ultrasound to target the appropriate region for depositing local anesthetics, guiding the needle to the intended region, and visualizing the infusion of anesthetics.
Ordering information: These items cannot be returned if used. Please consult with your sales representitive before purchasing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty