Would you like to log in again?
Digital Microscope Imager, 2 MP
Supplier: CELESTRON INTERNATIONAL INC SE
44423
470327-776EA 80.75 USD InStock
470327-776
Digital Microscope Imager, 2 MP
Microscopes
Capture images and video of your specimens via your computer.
  • 2 MP image sensor makes a traditional microscope into a digital microscope
  • Works with 22 and 30 mm diameter eyepieces
  • Connect the imager to your computer via USB 2.0 cable
  • New celestron S-viewer imaging software for Windows 7/8/10
  • Imager itself compatible with Mac OS photo booth and quick camera
Turn your traditional microscope (stereo and biological) into a high-resolution digital microscope using a 2 MP CMOS sensor digital microscope imager. Place the imager over the eyepiece of your microscope and stream the image to your computer via a USB cable. It's the perfect tool for hobbyists, teachers, students, medical labs, exhibits and more.
Use the included Windows software to capture images and video of your discoveries. Measure and annotate your specimens with built-in measurement and marking tools. S-Viewer software is only compatible with Windows (XP, Vista, Windows 7/8/10). Imager can be used directly as an external camera on Mac OS systems via photo booth or quick camera.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty