Would you like to log in again?
Accucold Med-Lab Performance Series Refrigerators
ARG3ML ARS6MLDR ARS12MLDR ARS1ML ARS15MLDR ARG1ML ARG18ML ARG6MLDR ARG8MLDR ARS18ML ARG15ML ARG12ML ARS8MLDR ARS6ML ARG8ML ARG12MLDR ARS8ML ARS3ML ARS15ML ARG15MLDR ARS12ML ARG6ML
470328-002EA 2984 USD InStock
470328-002 470328-004 470328-014 470328-006 470328-016 76636-496 470327-990 470327-994 76636-528 470327-988 470327-998 470327-986 76636-520 76636-498 76636-522 76636-500 76636-524 76636-502 470328-000 76636-526 76636-504 470328-010
Accucold Med-Lab Performance Series Refrigerators
Refrigerators
Designed and purpose-built to meet the demanding needs of scientific, life science, laboratory, and medical cold storage applications.
  • Solid and glass door refrigerators from 1 to 15 cubic feet
  • Built-in readout for current and high/low temperature
  • Audible and visual high/low temperature alarms
  • Optimized efficient performance for stable temperature
  • Eco-friendly natural refrigerants for a greener footprint
The all-refrigerator units come with an adjustable control range from 2 to 10 °C. The microprocessor temperature controller is externally located to minimize door openings, helping to protect stored contents from ambient temperature. The buffered temperature probe is encased in a glycol-filled bottle to better simulate temperature of the stored product, with an additional sensor that reads air temperature to ensure superior temperature control. The current and min/max temperature display is viewable in Celsius or Fahrenheit to the nearest tenth of a degree. The audible and visual temperature alarm sounds if the unit's interior goes out of range. Additional alarms include a power failure alert, door open alarm, and sensor failure alarm. This unit is equipped with remote alarm contacts in the rear and a ½" probe port on the right hand side to accept additional monitoring devices.
Inside, they utilizes adjustable cycle defrost operation to avoid temperature spikes. Optimized forced air cooling with two internal fans ensures excellent temperature stability and uniformity with rapid recovery after door openings. The adjustable plastic-coated wire shelves can be positioned at ½" increments to accommodate virtually any sized item. We include seven shelves, more than competitor products providing to maximize interior shelving storage area. This unit includes LED internal lighting with an on/off rocker switch. The interior is constructed from white powder coated metal to ensure added durability and better temperature retention.
They come in a white cabinet finish with a blue accented control panel and easy-grip handle. The user-reversible double pane tempered glass door provides a full view of the interior. These models include a factory-installed lock and ships with two keys for staff use. Powerful magnetic gaskets ensure a positive seal.
Designed with hydrocarbon refrigerant, the Pharm-Fac Performance Refrigerators provide an energy efficient solution to vaccine storage. These units are Pyxis, Omnicell, and AcuDose RX compatible.
Certifications: UL, ULC, ETL-S
Delivery information: They are delivered with 100% guarantee, 2 year-parts and labor and 5 year compressor warranty.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty