Would you like to log in again?
Hollow Acrylic Prism
Supplier: EISCO SCIENTIFIC LLC
PH0552F PH0552E
470328-124EA 14.4 USD InStock
470328-124 470328-126
Hollow Acrylic Prism
Educational Materials Physics Educational Materials Optics Learning Activities
Designed to study and demonstrate Snell's Law of Refraction.
  • Made with high quality acrylic plates
  • Edges and points are fully cemented
  • Includes stopper for leak-free use when filled
When referring to light or other waves passing through a boundary between two different isotropic media, Snell's law can be used to describe the relationship between the angles of incidence and refraction. Fill the hollow prism with a liquid of your choice. These are also useful when studying other principles of light manipulation, light rays, and light angles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty