Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acrylic Equilateral Prisms

Supplier: EISCO SCIENTIFIC LLC
PH0554AC PH0554DC PH0554CC PH0554SET PH0554BC
470328-134EA 4.65 USD InStock
470328-134 470328-136 470328-146 470328-130 470328-132
Acrylic Equilateral Prisms
Educational Materials Physics Educational Materials Optics Learning Activities
Designed for educational purposes and science classroom demonstrations of refraction principles and light physics.

  • Made of high quality acrylic
  • Refractive index almost identical to optical glass
  • Polished on all sides

High quality equilateral prisms are chip- and shatter-resistant, while having a refractive index very close to optical glass. They can produce a rainbow from a non-monochromatic light source such as sunlight. Acrylic is a softer material, so handle with care to prevent scratching.

Ordering information: The Acrylic Equilateral Prism Kit (470328-146) includes prisms 470328-132, 470328-134, and 470328-136.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty