Would you like to log in again?
Innovating Science® Distant Learning: Elephant Toothpaste
Supplier: ALDON CORP SE
Ward's®
IS1501 IS1501_U
470328-342KT 11.6 USD InStock
470328-342 470329-732
Innovating Science® Distant Learning: Elephant Toothpaste
Educational Classroom Kits and Activities
Learn about the effect a catalyst has on a chemical reaction.
  • Students will visualize the decomposition of hydrogen peroxide - as the reaction occurs, a large amount of foam is formed that flows up and out of the reaction bottle.
  • Aligned to NGSS Practices
  • Performance expectations: 2-PS1-4, 2-PS1-3, 5-PS1-1, 5-PS1-4
  • Disciplinary core ideas: PS1.A, PS1.B
  • Cross cutting concepts include cause and effect, energy and matter, scale proportion and quantity
This activity is a fun, safe way to teach students the effect a catalyst has on a chemical reaction. The decomposition of hydrogen peroxide can be visualized by mixing in a small amount of dish soap and adding yeast as a catalyst. As the reaction occurs, a large amount of foam is formed that flows up and out of the reaction bottle. This kit contains an Instruction Manual and enough materials to perform the demonstration twice.
Delivery information: Kit includes instructional manual and SDS booklet.
Caution: CHOKING HAZARD: Small parts. Not for Children under 3 years.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty