Would you like to log in again?
Innovating Science® Distant Learning: Paper Chromatography
Supplier: ALDON CORP SE
Ward's®
IS1502 IS1502_U
470328-344KT 17.95 USD InStock
470328-344 470329-734
Innovating Science® Distant Learning: Paper Chromatography
Educational Classroom Kits and Activities
Learn about chromatography, the oldest documented technique to separate chemical substances.
  • Students will perform paper chromatography on 3 individual dyes, along with a mixture of dyes, to determine the composition of the mixture.
  • Aligned to NGSS Practices
  • Performance expectations: MS-PS1-2, HS-PS1-3
  • Disciplinary core ideas: PS1.A, PS1.B
  • Cross cutting concepts include Patterns
  • Engineering practices include analyzing and interpreting data, planning and carrying out investigations
Chromatography is the oldest documented technique to separate chemical substances. In this activity, students will perform paper chromatography on three individual dyes, along with a mixture of dyes, to determine the composition of the mixture. Students will understand not only the components of a chromatography system, but also why different substances separate and move at different rates within the system. An Instruction Manual and enough materials for 2 groups are included.
Delivery information: Kit includes instructional manual and SDS booklet.
Caution:  This product can expose you to chemicals including Crystal Violet and Methanol/Methyl isobutyl ketone, which are known to the State of California to cause cancer and reproductive harm. For more information go to http://www.P65Warnings.ca.gov.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty