Would you like to log in again?
Ward's® DNA Simulation Lab Activity
Supplier: Avantor
WARD470023-864
470023-864KT 219.99 USD InStock
470023-864 470328-880
Ward's® DNA Simulation Lab Activity
Educational Classroom Kits and Activities
With this activity, students re-create the DNA molecule to better understand replication, transcription, and translation.
  • Pre- and post-laboratory assessments included
  • Model helps students visualize DNA molecule
  • Estimated lab time: 60 minutes
  • Colored beads represent DNA parts
  • Aligned to NGSS standards (MS/HS: LS1, LS3)
Students discover the intricacies of DNA when they build their own DNA models with pop beads. The beads are color coded to represent the phosphate/deoxyribose sugar backbone and the four nucleotide bases. The connectors between the pop beads represent hydrogen bonds.
Once the model is built, students can manipulate the molecule to visualize the processes of replication, transcription, and translation. They can also determine all possible codon combinations and the amino acids for which each codon specifies.
The kit is also available for purchase with a link to a NewPath digital lesson, which includes a variety of resources on the topic, including background information, illustrations and self-assessments.
Ordering information: This kit includes red pop beads (phosphate), white five-hole pop beads (deoxyribose sugar), yellow pop beads (adenine), orange pop beads (thymine), green pop beads (guanine), blue pop beads (cytosine), pink pop beads (uracil), and plastic connectors (hydrogen bonds).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty