Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Ward's® Prepared Agar Cubes
Supplier: RALPH CHARLTON MO
WARD470329-088 AGAR CUBES
470184-664KT 27.95 USD InStock
470184-664 470329-088
Ward's® Prepared Agar Cubes
Educational Materials Biology Educational Materials AP Biology Learning Activities
Make teaching diffusion easy. Exclusively from Ward's Science, we've pre-mixed, pre-poured, pre-measured and pre-cut these agar cubes to save you time and eliminate messy preparation in your lab. Just open the jar and begin using. Prepared to align with Ward's AP Biology Investigation 4: Diffusion and Osmosis, these cubes are also ideal for use on their own, or in any lab activity demonstrating the topic of diffusion. Agar cubes are prepared in three sizes (approximately 1cm, 2cm, and 3 cm). Phenolphthalein has been added to the cubes to exhibit a vivid fuchsia color during production and stored in a solution of sodium hydroxide. Just add an acid such as vinegar to demonstrate diffusion.
Each class size jar of agar cubes contains 18 cubes total and is composed of 6 each of the following sizes:
1 x 1 cm
2 x 2 cm
3 x 3 cm
Individual jars contain one of each, for a total of three cubes.
Ordering information: At times, supply may be limited due to high demand.  Please place your order approximately two weeks in advance.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty