Would you like to log in again?
Monocular Cordless LED Microscope, National
Supplier: NATIONAL OPTICAL & SCIENTIFIC SE
National®
131-RLED
470329-172CS 1535.2 USD InStock
470329-172
Monocular Cordless LED Microscope, National
Microscopes
Quality at an affordable price.
  • DIN 4x, 10x, and 40xR, objectives are achromatic, parfocalled, parcentered, and color-coded
  • 45° inclined viewing head rotates 360° for easy repositioning
  • Single 8 hours charge allows up to 50 hours of use
The microscope that sets the standard for high school instruction, now with the freedom of rechargeable batteries. Includes all the most commonly specified features, like rugged metal construction and locked-on parts. Superior DIN optical system and precision mechanical components assure parfocalled and parcentered optical alignment for sharp, unsurpassed viewing. Utilizes rechargeable batteries for bright, cool illumination from long-life LED bulbs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty