Would you like to log in again?
Dual Magnification Stereomicroscope, National
Supplier: NATIONAL OPTICAL & SCIENTIFIC SE
National®
452TBL-10
470329-174CS 1237.25 USD InStock
470329-174
Dual Magnification Stereomicroscope, National
Microscopes
Quality at an affordable price.
  • Eyepieces equipped with reticle holder/retainer to accept reticle with 23 mm Ext.Ø
  • 4-position light control: top only, bottom only, top and bottom, off
  • Fixed head
All the quality features and versatility of larger stereo microscopes, in a compact size only 12" tall. Ideal size for elementary and middle school, rugged enough for high school. Upright, unreversed image is designed for three-dimensional objects and dissecting. Choose low magnification for a wide field of view, then high magnification for close-up detail.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty