Would you like to log in again?
Innovating Science® DNA Extraction Lab Activity
Supplier: ALDON CORP SE
Innovating Science®
IS3002 IS1505
470343-470EA 48.5 USD InStock
470343-470 470329-580
Innovating Science® DNA Extraction Lab Activity
Educational Classroom Kits and Activities
Learn the history of the discovery of DNA and DNA structure, the nature of genetic inheritance, and the tole of DNA and proteins in genetic expression.
  • Students will isolate DNA from plant material
  • Aligned to NGSS Practices
  • Performance expectations: HS-LS1-1, HS-LS3-1, HS-LS3-2, HS-LS3-3
  • Disciplinary core ideas: LS1.A, LS3.A, LS3.B
  • Cross cutting concepts include structure and function, cause and effect, scale porportion and quantity, science is a human endeavor
  • Engineering practices include constructing explanations and designing solutions, asking questions and defining problems, analyzing and interpreting data, engaging in argument from evidence
In this lab you will learn the history of the discovery of DNA and DNA structure. Understand the nature of genetic inheritance and the role of DNA and proteins in genetic expression while using biological detergents, enzymes, and ethanol to isolate DNA from plant material. You need to supply the plant material.
Kit contains enough materials for 15 groups. Teacher's Manual and Student Study Guide copymasters are included.
Delivery information: Instructor's manual and SDS included.
Caution: This product can expose you to chemicals including Methanol/Methyl isobutyl ketone, which is known to the State of California to cause cancer and reproductive harm. For more information go to http://www.P65Warnings.ca.gov.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty