Would you like to log in again?
Innovating Science® Kidneys and blood filtration lab activity
Supplier: ALDON CORP SE
Innovating Science®
IS1506 IS3006
470329-582EA 19.2 USD InStock
470329-582 470343-476
Innovating Science® Kidneys and blood filtration lab activity
Educational Classroom Kits and Activities
Learn the role of the kidney in blood filtration and waste removal along with the many functional tasks performed by nephrons.
  • Students will create an artificial kidney model to filter simulated blood
  • Aligned to NGSS Practices
  • Performance expectations: MS-PS1-2, MS-LS1-3, HS-LS1-3
  • Disciplinary core ideas: PS1.A, PS1.B, LS1.A
  • Cross cutting concepts include patterns, systems and system models, stability and change
  • Engineering practices include analyzing and interpreting data, planning and carrying out investigations
Learn the role of the kidney in blood filtration and waste removal along with the many functional tasks performed by nephrons, as well as nephron structure. Students will create an artificial kidney model to filter simulated blood. This will allow them to visually determine if filtration of the simulated blood may or may not have occurred.
Chemically test the resulting filtrate to detect any possible waste material that may have been removed by the kidney. Kit contains enough materials for 15 groups. Teacher's Manual and Student Study Guide copymasters are included.
Delivery information: Instructor's manual and SDS included.
Caution: This product can expose you to chemicals including Benzidine-based dyes, which is known to the State of California to cause cancer. For more information go to http://www.P65Warnings.ca.gov.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty