Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sprint™ Clinical Centrifuges, 230 V

Supplier: Ward's

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

C5000-6H-E C3303-6HP-E
470329-986EA 609 USD InStock
470329-986 470329-990
Sprint™ Clinical Centrifuges, 230 V
Centrifuges Benchtop Centrifuges
These economical swing out rotor centrifuges provide fixed-speed or variable speed capabilities for all your blood and urine collection tubes up to 15 ml.

  • 230 V models, not for use in USA
  • 6 position swing-out rotor
  • Choose either fixed or variable speed
  • Adjustable timer

These clinical centrifuges are the ideal economical choice for all your blood, urine and cell separation needs. Both models offer a variable timer and digital display for settings. The basic model has a fixed speed, while the Plus model offers variable speed, in addition to a larger capacity tube (up to 15 ml, round or conical bottom), as well as increased speed, a safety imbalance detection system, momentary operation, and adjustable acceleration and deceleration levels. Both come with a 2 year warranty.

Certifications: UL listed.

Caution: 230 V models, not for use in USA. Corresponding models with voltage suitable for use in US (470329-984 and 470329-988) are also available, please see separate product page.

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty