Would you like to log in again?
Formaldehyde-Free Pig Hearts
Supplier: VWR International
Ward's®
140.1433.1
470330-138EA 13.75 USD InStock
470330-138
Formaldehyde-Free Pig Hearts
Specimens Preserved Specimens Preserved Animal Materials
Formaldehyde-free pig hearts, available in vacuum packs.
  • Mammalian anatomy
  • High quality preservation
To better serve the needs of all teaching environments and preferences, WARD’S offers this line as an alternative to our existing line of high-quality, safe preserved specimens. Absolutely no formaldehyde whatsoever is used in the fixing and preserving process for these specimens. Instead, our new solution used for preparation binds with the tissue as non-toxic, cross-linked proteins, making the solution truly 100% formaldehyde-free.
WARD’S offers the largest selection of specimens prepared with this alternative process, including the widest range of sizes and injection styles. In addition, the specimens can be used comfortably over extended periods of time.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty