Would you like to log in again?
Parafilm® M Box Top Dispenser
Supplier: Heathrow Scientific
Parafilm® M
120758W
470330-530CS 1049.9 USD InStock
470330-530
Parafilm® M Box Top Dispenser
Sealing Film Dispensers
Easy application with no mess.
  • Accommodates 2×50 mm or 1×100 mm Parafilm® M rolls
  • No need to remove rolls from packaging
  • Includes two safety blades
Simply place the dispenser on top of the Parafilm®M box and pull the sealing film through for quick dispensing. The built-in safety blade slides and cuts with no danger to fingertips, and eliminates the need for scissors.
Dispenser does not come with Parafilm® M rolls. Rolls are available separately.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty