Fresh frozen (unpreserved) dissection specimens shipped in coolers of ice directly to your classroom.
- Fresh frozen specimens
- Chemical free dissection
- Realistic experience
Fresh frozen dissection specimens allow you to experience dissection without the incorporation of chemicals for preservation.
Fresh vs. Frozen: Fresh specimens are only refrigerated; they are not frozen. Fresh frozen specimens are kept in freezers until shipped.
Ordering information: While fresh frozen specimens may be immediately available, some products may have a lead time of several weeks. Please place your orders ahead of time and allow extra time for delivery. Extra costs include coolers, packaging, and ice.
Delivery information: We suggest fresh frozen items are shipped on Monday, Tuesday, or Wednesday so they won't be held over the weekend.
Packaging: To maintain integrity, we recommend fresh frozen items be sent overnight or 2-day delivery and put immediately into a refrigerator or freezer upon arrival.
Caution: Storage Conditions : Cooler