Would you like to log in again?
Ward's® Pure Preserved Snakes
Supplier: VWR
Ward's®
WARD470330-658 WARD470314-626 WARD470314-692 WARD470330-660
470314-626PK 254.89 USD InStock
470314-626 470330-660 470330-658 470314-692
Ward's® Pure Preserved Snakes
Specimens Preserved Specimens Preserved Animal Materials
Ward's® PURE Preserved Specimens - the best dissection experience from start to finish.
  • Safest specimens
  • Low formalin odor
  • Best classroom experience
Ward's® PURE preserved specimens offer the best classroom experience because PURE specimens are safe. Students stay focused on learning and not on smells. Experience quality with true-to-life preservation, combined with artfully illustrated dissection guides, free with every purchase, to maximize value, lesson time and student engagement.
We guarantee you’ve never experienced dissection like this before. Go Pure.
Snakes are common reptiles used in dissection and to teach, among other things, vertebrate anatomy and comparative physiology. Students can explore the many vertebrae along their body in comparison to humans, who have just 32 vertebrae. Many snakes are camouflaged by color and pattern to blend into their surroundings and mimic their natural habitat. Garter snakes or other common non-poisonous species are supplied.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty