Would you like to log in again?
Nucleic Acid Testing for COVID-19
Supplier: Edvotek
123
470330-672EA 108.99 USD InStock
470330-672
Nucleic Acid Testing for COVID-19
Educational Materials Biology Educational Materials Genetics Learning Activities
Hands-on simulation of molecular testing for COVID-19.
  • Students learn about characteristics of Coronaviruses
  • Includes all required reagents
  • Use DNA Electrophoresis to analyze simulated samples
  • Activity can be completed in 45 minutes
SARS-CoV-2 is a novel coronavirus that has caused a worldwide outbreak of respiratory disease beginning in 2019.
In this simulated medical test, we will use electrophoresis to detect the presence of the SARS-CoV-2 virus in samples from patients with symptoms of COVID-19.
Additional items needed but not included: DNA electrophoresis equipment, white-light box, microwave or hot plate.
Ordering information: Kit will ship immediately upon ordering. Please call to specify a future shipping date.
Caution: Store DNA samples in the refrigerator. All other components may be stored at room temperature.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty