Would you like to log in again?
A-maize-ing Editing: Using CRISPR to Improve Crops
Supplier: Edvotek
210
470330-680EA 160.79 USD InStock
470330-680
A-maize-ing Editing: Using CRISPR to Improve Crops
Educational Materials Biology Educational Materials Genetics Learning Activities
Explore cutting edge biotechnology with this CRISPR simulation.
  • Hands-on CRISPR Simulation
  • Create Guide RNAs (gRNAs)
  • Use BLAST to verify gRNAs' specificity
  • Test your gRNAs using gel electrophoresis
  • Complete in approx. 1 hour 45 min
Students will assume the role of plant geneticists working to develop corn crops capable of surviving, and thriving, in a changing environment.
This experiment will allow students to develop an understanding of CRISPR-Cas9 applications in the laboratory, cleave DNA, and examine their results after gel electrophoresis.
Additional equipment needed but not included: DNA electrophoresis equipment, microwave or hot plate.
Ordering information: Kit will ship immediately upon ordering. Please call to place your order if you would like to specify a future date for shipment.
Caution: Some components require Freezer storage upon receipt.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty