Would you like to log in again?
Mother of All Experiments: Exploring Human Origins by PCR of Mitochondrial DNA
Supplier: Edvotek
332-FB 332
470330-700EA 129 USD InStock
470330-700 470330-702
Mother of All Experiments: Exploring Human Origins by PCR of Mitochondrial DNA
Educational Materials Biology Educational Materials Genetics Learning Activities
Let your students isolate, amplify and visualize their own Mitochondrial DNA.
  • Students isolate and analyze their own DNA
  • Total time (with stop-points) approximately 4 hours
  • Perishable component refills available
In this experiment, students will isolate their mitochondrial DNA and use the Polymerase Chain Reaction (PCR) to amplify two separate regions of the mitochondrial genome. Results are analyzed using agarose gel electrophoresis.
Time Required: Extraction - 50 min; PCR Setup - 10 min; PCR - 2 hours; Electrophoresis - 1 hour; Staining - 5 min to overnight.
Additional items needed but not included: Electrophoresis equipment, Thermal Cycler, 50 µl micropipettes; Light box, 2 water baths, microcentrifuge, microwave/hot plate.
Ordering information: Kit will ship immediately upon ordering. Please call to place your order if you would like to specify a future date for shipment.
Caution: Some components require Freezer storage upon receipt.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty