Would you like to log in again?
Learn to Code! Introduction to Python for Detecting Disease
Supplier: Edvotek
342
470330-704EA 57.15 USD InStock
470330-704
Learn to Code! Introduction to Python for Detecting Disease
Educational Materials Biology Educational Materials Human Biology Learning Activities
Introduce your students to biological data analysis.
  • Learn to code in computer programming language Python
  • No prior coding knowledge required
  • Master 15 fundamental coding skills
  • Create a program that analyzes DNA sequences and identifies mutations
  • Total time: 3 to 4 hr
Bioinformatics is a field dedicated to biological data analysis; an interface between computers and human health. Scientists use powerful computers to create simulations of biological activity and process large data sets, making large and complex data sets easy to understand and analyse.
In this experiment, students will learn how bioinformaticians process gene data, how they make sense of gene behavior, and what happens genotypically when a mutation occurs in a DNA sequence.
First, students will be introduced to the basics of the coding language Python, and will then use these skills to identify a SNP in a simulated patient's DNA.
Each purchase includes access to the coding program for a whole class and PDF instructions. Users only need a computer with internet access.
Ordering information: Kit will ship immediately upon ordering. Please call to place your order if you would like to specify a future date for shipment.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty