Would you like to log in again?
Discovering Quantitative PCR Amplification (qPCR) and Analysis
Supplier: Edvotek
380
470330-710EA 340.69 USD InStock
470330-710
Discovering Quantitative PCR Amplification (qPCR) and Analysis
Educational Materials Biology Educational Materials PCR Learning Activities
Get hands-on experience with the principles and practice of qPCR.
  • This kit must be used with a RT Thermal Cycler
  • Learn to perform an advanced molecular biology procedure used in labs worldwide
  • Quantify DNA concentration of 4 experimental samples using a standard curve
Quantitative PCR (qPCR or real-time PCR) simultaneously amplifies and detects targeted DNA allowing scientists to discover the starting amount of a specific DNA sequence in an experimental sample. This powerful and precise technology has become a cornerstone of modern genetics research, biological monitoring, and medical diagnostics.
For students, performing qPCR offers the challenge of performing a mathematically and technically advanced experiment, the opportunity to understand key molecular concepts through hands-on learning, and an essential skill for future research work.
In this specially adapted education qPCR experiment, students will quantify the DNA concentration of four experimental samples using a standard curve approach and then confirm the experiment’s specificity and accuracy through gel electrophoresis, melt curve analysis, and data analysis.
Time Required: qPCR Setup - 45 min; qPCR-90 min; Electrophoresis - 1 hr; Staining - 5 min to overnight, Analysis of results - 45 min.
Additional items needed but not included: qPCR Thermal Cycler, Microcentrifuge, 50 μl Micropipette, Electrophoresis equipment and light-box, Microwave or Hot Plate, Pipet Pump, Balance, Hot Gloves, Laboratory Gloves, Beaker, Distilled Water, Ice.
Ordering information: Kit will ship immediately upon ordering. Please call to place your order if you would like to specify a future date for shipment. Requires wet-ice shipment with overnight delivery.
Caution: Some components require Freezer storage upon receipt.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty