Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Advanced Magnetic Stirrers
Supplier: VWR
30802215
470330-736EA 479 USD InStock
470330-736
VWR® Advanced Magnetic Stirrers
Stirrers Magnetic Stirrers
These top-of-the-line units combine advanced digital features with high quality and reliability.
  • All models feature digital displays
  • Speed range: 60 to 1600 rpm (stirring models)
  • All models include a 5 year warranty
These high quality units feature an advanced digital display. Advanced electronics provide speed stability (±2%). The new design reduces benchtop footprint by over 20%.
The speed range is adjustable in 10 rpm increments, with equivalent readability. The enclosure is made of polycarbonate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty