Would you like to log in again?
Edvotek Battling Bacteria: Ecosystem Dynamics in a Petri Dish
Supplier: Edvotek
935
470331-014EA 154.2 USD InStock
470331-014
Edvotek Battling Bacteria: Ecosystem Dynamics in a Petri Dish
Educational Materials Biology Educational Materials Microbiology Learning Activities
Understand niches, biotic/abiotic factors, and species interactions.
  • Create and manipulate a microbial ecosystem
  • Investigate competition and test Gause's law
  • Explore how different species respond to stress and environmental change
  • Experiment requires one week to complete
In this experiment students create their own microbial ecosystems and investigate the dynamics of competition and abiotic or biotic change on community composition. By using bacteria with short generation times students can observe, test, and collect their own data on key ecological phenomena in a week.
Additional items needed but not included: Micropipettes, incubator, centrifuge, marking pens, Bunsen burner, hot plate or microwave, and hot gloves.
Ordering information: Kit will ship immediately upon ordering. Please call to specify a future shipping date.
Caution: Some components require refrigerator and freezer storage.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty