Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Metalware Rod and Clamp Set, Eisco Scientific

Supplier: Eisco Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

MTST1VWR
470331-186EA 55.95 USD InStock
470331-186
Metalware Rod and Clamp Set, Eisco Scientific
Support Stands
A basic, three piece rod and clamp set useful in all science laboratories.

  • The 8" long by 5" wide base weighs 1.75 lbs, providing superb stability for experiments and setups
  • Base features a tapped hole, which fits the included rod, and can accommodate other rods with a 10×1.5 mm metric thread
  • The included rod measures 23.6" tall and is 10 mm in Ø
  • The 4 finger cork lined clamp is 10" long

Lab stands are used in every area of science - especially chemistry, for holding and manipulating an experimental apparatus. The durable and sturdy construction of this set makes it an excellent choice for science classrooms and basic professional laboratories.

The retort stand is made of corrosion and scaling resistant, powder coated pressed steel, and features a tapped hole, suitable for connecting the included rod, or any other rods with a 10×1.5 mm metric thread. Included rod is made of stainless steel and measures 23.6" tall, 10 mm in Ø and features a 10×1.5 mm metric thread. Clamp is four-pronged with a cork lining and is 10" long.

Ordering information: Includes retort base made of corrosion resistant, powder coated steel, a stainless steel rod featuring a 10×1.5 mm metric thread and a four-pronged cork-lined clamp.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty