Would you like to log in again?
Motic Swift Line M2651C Monocular Cordless LED Microscope
Supplier: Motic
Ward's®
M2651C
470331-190EA 344.5 USD InStock
470331-190
Motic Swift Line M2651C Monocular Cordless LED Microscope
Microscopes
Top quality optics, ideal for school use.
  • Achromatic objectives
  • Large stage
The M2651C is ideal for a wide variety of educational applications. This series has locked-on one touch stage clips, eyepiece, objectives and easy to use coaxial focusing system. M2651C is rechargeable, with cool and efficient dimmable LED illumination.
Objectives: 4× (0.10 N.A.), 10× (0.25 N.A.) and 40×R (0.65 N.A.) achromatic, DIN standard
Delivery information: Unit shipped with American plug.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty