Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Standard / VWR® Advanced Standard and Advanced Compound Microscopes
Supplier: VWR
VWR®
L500-2 L1000-1 L500-1
470331-636EA 590.75 USD InStock
470331-636 470331-634 470331-632
VWR® Standard / VWR® Advanced Standard and Advanced Compound Microscopes
Microscopes
Quality microscopes at an affordable price.
  • Wide-field 10X eyepiece with pointer
  • Parfocal and parcentered optics
  • Separate coarse and fine adjustment controls
  • Three-year manufacturer's warranty
These durable microscopes feature all-metal bodies and all-glass-coated optics. They provide crisp, clear images allowing students to view more details and minute structures.
Delivery information: All microscopes include a manual and a dust cover.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty