Would you like to log in again?
Supertek® Brewster's Angle Experiment
Supplier: Shiv Dial Sud & Sons
Supertek®
BAE1-C
470331-752EA 983.2 USD InStock
470331-752
Supertek® Brewster's Angle Experiment
Educational Materials Physics Educational Materials Optics Learning Activities
Measure Brewster’s angle for a given dielectric dispersive medium.
  • Compact rotating table makes the apparatus extremely smooth to operate
  • Sensitive light sensor (1 lux) enables the measurement of light intensity at various angles of reflection
When un-polarized light is incident on the surface of a dielectric (such as a glass), at a certain angle of incidence the reflected light is completely plane-polarized. This phenomenon was discovered by Sir David Brewster and, thus, the specific angle is called Brewster’s angle or polarization angle. Also, from the experiment, it can be confirmed that the reflected ray and the refracted ray are 90° apart when the incident angle is set at Brewster’s angle.
Delivery information: Kit includes optical bench set 0.8 m, light source holder, converging lens holder, adjustable collimating slit holder, rotating table assembly, analyzer square, light sensor holder, power supply for light source, glass block, data processor.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty