Would you like to log in again?
Supertek® Biot-Savart's Law Experiment
Supplier: Shiv Dial Sud & Sons
Supertek®
BSL1-C
470331-754EA 708 USD InStock
470331-754
Supertek® Biot-Savart's Law Experiment
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Measure the magnetic fields produced by straight and coiled conductors.
  • Multiple coil variants allow students to compare current, number of turns, radius
  • 2-axis Teslameter allows students to measure both axial and tangential magnetic fields
Electric currents generate magnetic fields. Biot–Savart law is an equation describing the magnetic field generated by a constant electric current. It relates the magnetic field to the magnitude, direction, length, and proximity of the electric current. In this experiment, we study the magnetic field characteristics in the straight conductors and different types of circular coils.
Delivery information: Kit includes optical bench set 0.4 m, solenoid with variable turns, solenoid with variable lengths, solenoid with different winding density (set), straight conductor holder, Teslameter with axial and tangential hall probe, power supply.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty