Would you like to log in again?
Eisco LabGlass® Boiling Flasks, Flat Bottom with Threaded Joint
Supplier: EISCO SCIENTIFIC LLC
CH01004E CH01004G CH01004D CH01004F CH01004B CH01004J CH01004I CH01004A CH01004H CH01004C
470331-804EA 16.9 USD InStock
470331-804 470331-802 470331-808 470331-792 470331-806 470331-796 470331-794 470331-800 470331-810 470331-798
Eisco LabGlass® Boiling Flasks, Flat Bottom with Threaded Joint
Flasks Flat Bottom Flasks
High quality flat bottom boiling flasks for chemistry classrooms and laboratories.
  • Made of high quality borosilicate glass 3.3
  • Ground threaded joint for secure fit with other glassware
  • Flat bottom design
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
  • Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C
Eisco manufactures all LabGlass products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty