Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Eisco LabGlass® Distillation Flasks, 2 Neck with Threaded Joint
Supplier: Eisco Scientific
CH01008A CH01008D CH01008F CH01008E CH01008C CH01008B
470331-836EA 36.7 USD InStock
470331-836 470331-834 470331-838 470331-842 470331-832 470331-840
Eisco LabGlass® Distillation Flasks, 2 Neck with Threaded Joint
Flasks Distillation Flasks
High quality distillation flask for chemistry classrooms and laboratories.
  • Made of high quality borosilicate glass 3.3
  • Ground threaded joints for secure fit with other glassware
  • Two neck design
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
Heat tolerances: strain point 515 °C; annealing point 565 °C and softening point 820 °C.
Eisco manufactures all LabGlass products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty