Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Eisco LabGlass® Distillation Flasks, 3 Necks with Threaded Joint
Supplier: Eisco Scientific
CH01010D CH01010C CH01010B CH01010F CH01010A CH01010E
470331-846EA 52.8 USD InStock
470331-846 470331-848 470331-852 470331-854 470331-844 470331-850
Eisco LabGlass® Distillation Flasks, 3 Necks with Threaded Joint
Flasks Distillation Flasks
High quality distillation flask for chemistry classrooms and laboratories.
  • Made of high quality borosilicate glass 3.3
  • Ground threaded joints for secure fit with other glassware
  • 3 neck design (angled)
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
  • Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C
Eisco manufactures all LabGlass products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty