Would you like to log in again?
Eisco LabGlass® Dropping Funnels with Threaded Joint
Supplier: EISCO SCIENTIFIC LLC
CH01024E CH01024C CH01024B CH01024A CH01024D
470331-904EA 53.35 USD InStock
470331-904 470331-900 470331-902 470331-896 470331-898
Eisco LabGlass® Dropping Funnels with Threaded Joint
Funnels Dropping Funnels
High quality dropping funnel for Chemistry classroom or laboratories.
  • Made of high quality borosilicate glass 3.3
  • Cylindrical design with PTFE key stopcock
  • Interchangeable, ground threaded joint for secure fit with other glassware
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
  • Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C
Eisco manufactures all LabGlass products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty