Would you like to log in again?
Eisco LabGlass® Reagent Bottles with Hexagonal Glass Stopper, Wide Mouth
Supplier: EISCO SCIENTIFIC LLC
CH0163F CH0163B CH0163C CH0163A CH0163E CH0163D
470331-938EA 13.65 USD InStock
470331-938 470331-944 470331-934 470331-936 470331-940 470331-942
Eisco LabGlass® Reagent Bottles with Hexagonal Glass Stopper, Wide Mouth
Bottles Glass Bottles
High-quality glass reagent bottle useful for storing chemicals in classrooms or laboratories.
  • Made of high-quality borosilicate glass 3.3
  • Interchangeable, ground glass stopper
  • Ungraduated
  • Wide mouth design
  • Fully autoclavable
  • Superior chemical durability
  • Amber colored glassware protects light-sesitive chemicals
Low temperature gradient - can withstand high temperatures and thermal shocks. Heat tolerances: Strain point 515 °C; annealing point 565 °C; softening point 820 °C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty