Would you like to log in again?
Eisco LabGlass® Crow Receivers, Class B
Supplier: EISCO SCIENTIFIC LLC
CH0351B CH0351A CH0351C
470331-956EA 18 USD InStock
470331-956 470331-958 470331-960
Eisco LabGlass® Crow Receivers, Class B
Distillation Components Distillation Receivers
High quality glass crow reciever useful for oil distilation and tar tests.
  • Made of high quality borosilicate glass 3.3
  • Features round base and spout
  • Chemical resistant blue graduations
  • Class B
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
  • Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C
Eisco manufactures all LabGlass products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty