Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Eisco LabGlass® Boiling Flasks, Round Bottom, Narrow Neck
Supplier: EISCO SCIENTIFIC LLC
CH0402G CH0402F CH0402C CH0402B CH0402D CH0402E CH0402A
470331-966EA 4.55 USD InStock
470331-966 470331-968 470331-962 470331-974 470331-964 470331-970 470331-972
Eisco LabGlass® Boiling Flasks, Round Bottom, Narrow Neck
Flasks Round Bottom Flasks
High quality round bottom boiling flasks with a narrow neck design.
  • Made of high quality borosilicate glass 3.3
  • Narrow neck design with beaded rim
  • Round bottom
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
Heat tolerances: strain point 515 °C; annealing point 565 °C and softening point 820 °C.
Eisco manufactures all LabGlass products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty