Would you like to log in again?
Eisco LabGlass® Boiling Flasks, Flat Bottom, Narrow Neck
Supplier: EISCO SCIENTIFIC LLC
CH0410B CH0410H CH0410D CH0410A CH0410G CH0410F CH0410K CH0410E CH0410C
470332-040EA 5.65 USD InStock
470332-040 470332-032 470332-042 470332-034 470332-044 470332-036 470332-046 470332-038 470332-048
Eisco LabGlass® Boiling Flasks, Flat Bottom, Narrow Neck
Flasks Flat Bottom Flasks
High quality flat bottom boiling flasks with a narrow neck design.
  • Made of high quality borosilicate glass 3.3
  • Narrow neck design with beaded rim
  • Flat bottom
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
  • Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C
Eisco manufactures all LabGlass products using state-of-the-art equipment and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty