Would you like to log in again?
Eisco LabGlass® Boiling Flasks, Flat Bottom, Interchangeable Joint
Supplier: EISCO SCIENTIFIC LLC
CH0412B02 CH0412H CH0412B CH0412F CH0412L CH0412B01 CH0412G CH0412E CH0412K CH0412D CH0412J CH0412I CH0412C CH0412I01 CH0412A
470332-090EA 25.15 USD InStock
470332-090 470332-070 470332-080 470332-072 470332-082 470332-074 470332-084 470332-062 470332-076 470332-064 470332-086 470332-078 470332-066 470332-088 470332-068
Eisco LabGlass® Boiling Flasks, Flat Bottom, Interchangeable Joint
Flasks Flat Bottom Flasks
High quality flat bottom boiling flasks with ground, interchangeable joint.
  • Made of high quality borosilicate glass 3.3
  • Interchangeable, ground joint
  • Stopper not included
  • Flat bottom
  • Autoclavable
  • Superior chemical durability
  • Low temperature gradient - can withstand high temperatures and thermal shocks
  • Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C
Eisco manufactures all LabGlass products using state-of-the-art equipment and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty